Skip to Content
Merck
All Photos(1)

Key Documents

536288

Sigma-Aldrich

2-Fluorobenzoic hydrazide

97%

Synonym(s):

2 Flurobenzhydrazide

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
€76.50

€76.50


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
€76.50

About This Item

Linear Formula:
FC6H4CONHNH2
CAS Number:
Molecular Weight:
154.14
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€76.50


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

mp

70-74 °C (lit.)

functional group

amine
fluoro
hydrazine

SMILES string

NNC(=O)c1ccccc1F

InChI

1S/C7H7FN2O/c8-6-4-2-1-3-5(6)7(11)10-9/h1-4H,9H2,(H,10,11)

InChI key

YJCCKQQVXNNAAR-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shao-Song Qian et al.
Acta chimica Slovenica, 62(4), 828-833 (2015-12-19)
Reaction of [VO(acac)(2)] (where acac = acetylacetonate), benzohydroxamic acid (Hbha), and two similar aroylhydrazone ligands in methanol produced two benzohydroxamate-coordinated mononuclear vanadium(V) oxo complexes with general formula [VOL(bha)], where L = L(1) = N'-(5-bromo-2-hydroxybenzylidene)-2-fluorobenzohydrazide (H(2)L(1)), and L = L(2) =

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service