Skip to Content
Merck
All Photos(1)

Key Documents

534781

Sigma-Aldrich

Amano Lipase A from Aspergillus niger

≥120,000 U/g

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
€40.40

€40.40


Estimated to ship on03 April 2025



Select a Size

Change View
10 G
€40.40

About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.22

€40.40


Estimated to ship on03 April 2025


Quality Level

specific activity

≥120,000 U/g, pH 6.5, 45 °C (Optimum pH and temperature)
≥120,000 U/g

storage temp.

2-8°C

InChI

1S/C11H9N3O2.Na/c15-8-4-5-9(10(16)7-8)13-14-11-3-1-2-6-12-11;/h1-7,16H,(H,12,14);/q;+1/b13-9-;

InChI key

QWZUIMCIEOCSJF-CHHCPSLASA-N

Related Categories

Application

Enzyme used for:
  • Chemoselective O-deacetylation reactions
  • Hydrophilic ionic liquid-supported epoxidation reactions
  • Regioselective hydrolysis reactions

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nan Luo et al.
Molecules (Basel, Switzerland), 26(5) (2021-04-04)
This study investigated the effect of gel structure on the digestion of heat-set whey protein emulsion gels containing capsaicinoids (CAP), including the bioaccessibility of CAP. Upon heat treatment at 90 °C, whey protein emulsion gels containing CAP (10 wt% whey
Jon Alberdi-Cedeño et al.
Antioxidants (Basel, Switzerland), 9(3) (2020-03-22)
The aim of this study is the analysis of the in vitro digestion of corn oil, and of the effect of its enrichment with three levels of gamma- and alpha-tocopherol, by using, for the first time, 1H nuclear magnetic resonance
Jon Alberdi-Cedeño et al.
Antioxidants (Basel, Switzerland), 9(6) (2020-06-25)
The changes provoked by in vitro digestion in the lipids of olive oil enriched or not with different phenolic compounds were studied by proton nuclear magnetic resonance (1H NMR) and solid phase microextraction followed by gas chromatography/mass spectrometry (SPME-GC/MS). These

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service