Skip to Content
Merck
All Photos(1)

Key Documents

534560

Sigma-Aldrich

(S)-(+)-2-Amino-1-hexanol

97%

Synonym(s):

L-Norleucinol

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€155.00

€155.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€155.00

About This Item

Linear Formula:
CH3(CH2)3CH(NH2)CH2OH
CAS Number:
Molecular Weight:
117.19
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

€155.00


Check Cart for Availability

Request a Bulk Order

Assay

97%

optical activity

[α]20/D +14°, c = 1 in chloroform

bp

216-218 °C (lit.)

mp

35-40 °C (lit.)

functional group

amine
hydroxyl

SMILES string

CCCC[C@H](N)CO

InChI

1S/C6H15NO/c1-2-3-4-6(7)5-8/h6,8H,2-5,7H2,1H3/t6-/m0/s1

InChI key

DPEOTCPCYHSVTC-LURJTMIESA-N

Application

(S)-(+)-2-Amino-1-hexanol can be used in one of the key synthetic steps for the synthesis of house dust mite (HDM) peptidase allergen Der p 1 inhibitors.[1][2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

211.0 °F

Flash Point(C)

99.44 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The discovery of potent, selective, and reversible inhibitors of the house dust mite peptidase allergen Der p 1: an innovative approach to the treatment of allergic asthma
Newton GK, et al.
Journal of Medicinal Chemistry, 57(22), 9447-9462 (2014)
Allergen Delivery Inhibitors: Characterisation of Potent and Selective Inhibitors of Der p 1 and Their Attenuation of Airway Responses to House Dust Mite Allergens
Zhang J, et al.
International Journal of Molecular Sciences, 19(10), 3166-3166 (2018)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service