Skip to Content
Merck
All Photos(1)

Key Documents

533653

Sigma-Aldrich

5-Methylhydantoin

97%

Synonym(s):

5-Methyl-2,4-imidazolidinedione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H6N2O2
CAS Number:
Molecular Weight:
114.10
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

148-152 °C (lit.)

SMILES string

CC1NC(=O)NC1=O

InChI

1S/C4H6N2O2/c1-2-3(7)6-4(8)5-2/h2H,1H3,(H2,5,6,7,8)

InChI key

VMAQYKGITHDWKL-UHFFFAOYSA-N

Application

Reactant for:
Preparation of N-chlorohydantoins
The Mitsunobu reaction
Pseudomonas putida strain allowing conversion into L-amino acids
D-hydantoinase from Agrobacteria tumefaceins
Eznymatic racemization

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B A Nogueira et al.
The journal of physical chemistry. A, 124(31), 6303-6318 (2020-06-10)
The conformational space of 5-acetic acid hydantoin {5AAH; [2-(2,5-dioxoimidazolidin-4-yl)acetic acid]} was investigated by quantum chemical calculations performed at the DFT(B3LYP)/6-311++G(d,p) level of theory. A total of 13 conformers were located in the potential energy surface of the molecule, six of
[Studies on the chemical constituents of Pueraria lobata].
M H Chen
Zhong yao tong bao (Beijing, China : 1981), 10(6), 34-36 (1985-06-01)
M L Niku-Paavola et al.
Journal of applied microbiology, 86(1), 29-35 (1999-04-13)
New types of antimicrobial compounds were identified in the culture filtrate of Lactobacillus plantarum VTT E-78076. Activity was detected in the low molecular mass fraction separated by gel chromatography. This fraction totally inhibited the growth of the Gram-negative test organism
C L Soong et al.
FEMS microbiology letters, 158(1), 51-55 (1998-02-07)
Cyclic imide-transforming activity was found to be widely distributed in bacteria, yeast and molds. This activity was not correlated with cyclic ureide-transforming activity in bacteria, but there was some correlation in yeast and molds. These two activities are probably catalyzed
Anupama S Engineer et al.
Enzyme and microbial technology, 138, 109554-109554 (2020-06-13)
Overexpression of a novel hydantoinase (hyuH) from P. aeruginosa (MCM B-887) in E. coli yielded optically pure carbamoyl amino acids. The use of optically pure carbamoyl amino acids as substrates facilitates the synthesis of non-proteinogenic amino acids. The enzyme hyuH

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service