Skip to Content
Merck
All Photos(1)

Key Documents

533475

Sigma-Aldrich

(2-Chloro-5-pyridyl)methylzinc chloride solution

0.5 M in THF

Sign Into View Organizational & Contract Pricing

Select a Size

50 ML
€394.00

€394.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
50 ML
€394.00

About This Item

Empirical Formula (Hill Notation):
C6H5Cl2NZn
CAS Number:
Molecular Weight:
227.41
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

€394.00


Estimated to ship on30 May 2025


Request a Bulk Order

concentration

0.5 M in THF

bp

65 °C

density

0.992 g/mL at 25 °C

functional group

chloro

storage temp.

2-8°C

SMILES string

Cl[Zn]Cc1ccc(Cl)nc1

InChI

1S/C6H5ClN.ClH.Zn/c1-5-2-3-6(7)8-4-5;;/h2-4H,1H2;1H;/q;;+1/p-1

InChI key

LZKBDZYNYMRVHH-UHFFFAOYSA-M

Legal Information

Product of Rieke Metals, Inc.
Rieke is a registered trademark of Rieke Metals, Inc.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

1.0 °F - closed cup

Flash Point(C)

-17.2 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service