Skip to Content
Merck
All Photos(1)

Documents

533351

Sigma-Aldrich

2,6-Dichlorobenzylzinc chloride solution

0.5 M in THF

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H3CH2ZnCl
CAS Number:
Molecular Weight:
260.86
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

concentration

0.5 M in THF

bp

65 °C

density

0.991 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

Cl[Zn]Cc1c(Cl)cccc1Cl

InChI

1S/C7H5Cl2.ClH.Zn/c1-5-6(8)3-2-4-7(5)9;;/h2-4H,1H2;1H;/q;;+1/p-1

InChI key

NEPYXTRQHMZQAE-UHFFFAOYSA-M

Related Categories

Application

2,6-Dichlorobenzylzinc chloride can be used as a substrate in the palladium-catalyzed synthesis of 1-phenyl-2,5-hexanedione derivatives using methyl vinyl ketone and carbon monoxide.

Legal Information

Product of Rieke Metals, Inc.
Rieke is a registered trademark of Rieke Metals, Inc.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

1.0 °F - closed cup

Flash Point(C)

-17.2 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pd (0)-catalyzed conjugate addition of benzylzinc chlorides to α, β-enones in an atmosphere of carbon monoxide: Preparation of 1, 4-diketones
Yuguchi M, et al.
The Journal of Organic Chemistry, 69(3), 908-914 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service