Skip to Content
Merck
All Photos(1)

Key Documents

532347

Sigma-Aldrich

1,2:5,6-Di-O-isopropylidene-D-chiro-inositol

98%

Synonym(s):

1,2:5,6-Bis-O-(1-methylethylidene)-3-methyl-1D-chiro-inositol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H22O6
CAS Number:
Molecular Weight:
274.31
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

98%

optical activity

[α]20/D −42.5°, c = 2 in chloroform

mp

105-108 °C (lit.)

SMILES string

CO[C@H]1[C@H](O)[C@H]2OC(C)(C)O[C@H]2[C@@H]3OC(C)(C)O[C@H]13

InChI

1S/C13H22O6/c1-12(2)16-8-6(14)7(15-5)9-11(10(8)18-12)19-13(3,4)17-9/h6-11,14H,1-5H3/t6-,7-,8+,9+,10+,11+/m0/s1

InChI key

IPNUFAHKROGNMZ-HGQQYVGVSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Khurshid A Bhat et al.
Bioorganic & medicinal chemistry letters, 19(7), 1939-1943 (2009-03-10)
Semi-synthetic analogs of pinitol were subjected to screening by determining TNF-alpha expression in human neutrophils using flowcytometry. Among the tested compounds, three derivatives displayed more than 50% inhibition of TNF-alpha cytokine secretion in LPS induced stimulated neutrophils and can be

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service