Skip to Content
Merck
All Photos(1)

Documents

531421

Sigma-Aldrich

Trimethylsilyl 2,2-difluoro-2-(fluorosulfonyl)acetate

Synonym(s):

Trimethylsilyl (fluorosulfonyl)difluoroacetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FSO2CF2CO2Si(CH3)3
CAS Number:
Molecular Weight:
250.27
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

reaction suitability

reaction type: C-C Bond Formation

refractive index

n20/D 1.367 (lit.)

bp

156 °C (lit.)

density

1.27 g/mL at 25 °C (lit.)

SMILES string

C[Si](C)(C)OC(=O)C(F)(F)S(F)(=O)=O

InChI

1S/C5H9F3O4SSi/c1-14(2,3)12-4(9)5(6,7)13(8,10)11/h1-3H3

InChI key

XHVSCKNABCCCAC-UHFFFAOYSA-N

Application

  • Fluoroalkylation agent

Reactant for:
  • Preparation of difluorocyclopropenes
  • Preparation of difluorocarbene reagents

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

80.1 °F - closed cup

Flash Point(C)

26.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service