Skip to Content
Merck
All Photos(2)

Documents

526452

Sigma-Aldrich

2,4,5-Trimethylbenzaldehyde

90%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3C6H2CHO
CAS Number:
Molecular Weight:
148.20
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

90%

impurities

7% 5-bromo-1,2,4-trimethylbenzene

mp

41-46 °C (lit.)

SMILES string

[H]C(=O)c1cc(C)c(C)cc1C

InChI

1S/C10H12O/c1-7-4-9(3)10(6-11)5-8(7)2/h4-6H,1-3H3

InChI key

LROJZZICACKNJL-UHFFFAOYSA-N

General description

Dipole moments of 2,4,5-trimethylbenzaldehyde (TMB) isolated in a durene host crystal have been reported to be 1.65 ± 0.09D (difference between the ground state and the lowest singlet) and 1.05 ± 0.06D (difference between the ground state and triplet excited states). TMB has been identified as one of the major volatile constituent present in the leave extracts of Eryngium foetidum L. Based on the phosphorescence excitation and Stark spectral data, it has been observed that a change in temperature and electric field causes a change in the dipole moment of the compound.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H J Fang et al.
Yao xue xue bao = Acta pharmaceutica Sinica, 25(7), 534-537 (1990-01-01)
Essential oil from the rhizome of Cyclorhiza waltonii was obtained by steam distillation. The chemical components of the oil were examined by means of capillary gas chromatography and gas chromatography-mass selective detector with HP-5 and Carbowax 20M stationary phases. Fifteen
Composition of the leaf oil of Eryngium foetidum L. from Cuba.
Pino JA, et al.
J. Essent. Oil Res., 9(4), 467-468 (1997)
Triplet state Stark spectroscopy above the phosphorescence origin of 2, 4, 5-trimethylbenzaldehyde.
Winkler IC and Hanson DM.
J. Chem. Phys. , 76(12), 5739-5745 (1982)
Dipole moments of the lowest excited singlet and triplet states of 2, 4, 5-trimethylbenzaldehyde in a durene host crystal.
Sheng SJ and Hanson DM.
J. Chem. Phys. , 60(2), 368-374 (1974)
On the ?dual phosphorescence? from 2, 4, 5-trimethylbenzaldehyde in crystalline durene.
Moehle W and Vala M.
Chemical Physics Letters, 41(1), 149-153 (1976)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service