Skip to Content
Merck
All Photos(2)

Key Documents

524891

Sigma-Aldrich

1-(Triphenylmethyl)imidazole

97%

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€589.00

€589.00


Estimated to ship on12 April 2025


Request a Bulk Order

Select a Size

Change View
25 G
€589.00

About This Item

Empirical Formula (Hill Notation):
C22H18N2
CAS Number:
Molecular Weight:
310.39
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

€589.00


Estimated to ship on12 April 2025


Request a Bulk Order

Assay

97%

mp

220-224 °C (lit.)

functional group

phenyl

SMILES string

c1ccc(cc1)C(c2ccccc2)(c3ccccc3)n4ccnc4

InChI

1S/C22H18N2/c1-4-10-19(11-5-1)22(24-17-16-23-18-24,20-12-6-2-7-13-20)21-14-8-3-9-15-21/h1-18H

InChI key

NPZDCTUDQYGYQD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-(Triphenylmethyl)imidazole is an imidazole derivative.

Application

1-(Triphenylmethyl)imidazole may be used in the synthesis of 1,2,5-trisubstituted imidazole derivatives.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

2, 5-Dilithiation of N-protected imidazoles. Syntheses of 2, 5-disubstituted derivatives of 1-methoxymethyl-, 1-triphenylmethyl-, and 1-(N, N-dimethylsulphonamido)-imidazole.
Chadwick DJ and Ngochindo RI.
Journal of the Chemical Society. Perkin Transactions 1, 481-486 (1984)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service