Skip to Content
Merck
All Photos(1)

Key Documents

524778

Sigma-Aldrich

5-Methyl-2-furonitrile

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5NO
CAS Number:
Molecular Weight:
107.11
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay:
97%

Assay

97%

refractive index

n20/D 1.484 (lit.)

bp

173-176 °C (lit.)

density

1.02 g/mL at 25 °C (lit.)

SMILES string

Cc1ccc(o1)C#N

InChI

1S/C6H5NO/c1-5-2-3-6(4-7)8-5/h2-3H,1H3

InChI key

XGCRBVWSFYTMEC-UHFFFAOYSA-N

General description

5-Methyl-2-furonitrile is formed on reacting α-furfuryl chloride with strong aqueous potassium cyanide.[1][2]

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

138.9 °F - closed cup

Flash Point(C)

59.4 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The preparation of 2-Furanacetic acid.
Plucker III J and Amstutz ED.
Journal of the American Chemical Society, 62(6), 1512-1513 (1940)
Reactions in the β-Furan Series?Synthesis and Reactions of 3-Chloromethylfuran and Some Related Compounds.
Sherman E and Amstutz ED.
Journal of the American Chemical Society, 72(5), 2195- 2199 (1950)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service