Skip to Content
Merck
All Photos(1)

Key Documents

524204

Sigma-Aldrich

2,5-Dichloro-4-nitroaniline

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H2(NO2)NH2
CAS Number:
Molecular Weight:
207.01
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

154-158 °C (lit.)

functional group

chloro
nitro

SMILES string

Nc1cc(Cl)c(cc1Cl)[N+]([O-])=O

InChI

1S/C6H4Cl2N2O2/c7-3-2-6(10(11)12)4(8)1-5(3)9/h1-2H,9H2

InChI key

JBXZCPXEYAEMJS-UHFFFAOYSA-N

General description

2,5-Dichloro-4-nitroaniline is primary aniline indicator. Its pKa in sulfonate and water are -2.12 and -1.78, respectively.

Application

2,5-Dichloro-4-nitroaniline may be employed as an indicator for the estimation of acidity of strong acids.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Chronic 2 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shashi Kant Shukla et al.
The journal of physical chemistry. B, 117(8), 2456-2465 (2013-02-05)
Acidity functions were used to express the ability of a solvent/solution to donate/accept a proton to a solute. The present work accounts for the acidity determination of HCOOH, CH3COOH, and CH3CH2COOH in the alkylimidazolium-based protic ionic liquids (PILs), incorporated with
A redetermination and extension of the H 0 scale of acidity in aqueous perchloric acid.
Yates K and Wai H.
Journal of the American Chemical Society null
Solvent Effects in Organic Chemistry. II. Sulfolane1--A Weakly Basic Aprotic Solvent of High Dielectric Constant2.
Arnett EM and Douty CF.
Journal of the American Chemical Society, 86(3), 409-412 (1964)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service