Skip to Content
Merck
All Photos(1)

Key Documents

522937

Sigma-Aldrich

4-Amino-2-chloropyridine

97%

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
€129.00

€129.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
€129.00

About This Item

Empirical Formula (Hill Notation):
C5H5ClN2
CAS Number:
Molecular Weight:
128.56
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€129.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

mp

90-94 °C (lit.)

functional group

chloro

SMILES string

Nc1ccnc(Cl)c1

InChI

1S/C5H5ClN2/c6-5-3-4(7)1-2-8-5/h1-3H,(H2,7,8)

InChI key

BLBDTBCGPHPIJK-UHFFFAOYSA-N

General description

4-Amino-2-chloropyridine is a chloroaminoheterocyclic compound. It readily undergoes Suzuki-Miyaura coupling with phenylboronic acid.[1]

Application

4-Amino-2-chloropyridine may be used in the preparation of:
  • 3-deazacytosine[2]
  • 7-substituted 1-alky-1,4-dihydro-4-oxo-1,6-naphthyridine-3-carboxylic acids[3]
  • 1,6-naphthyridines[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation and biological activity of various 3-deazapyrimidines and related nucleosides.
A Bloch et al.
Journal of medicinal chemistry, 16(3), 294-297 (1973-03-01)
Pyridone-carboxylic Acids as Antibacterial Agents. I. Synthesis and Antibacterial Activity of 1-Alkyl-1, 4-dihydro-4-oxo-1, 8-and 1, 6-naphthyridine-3-carboxylic Acids.
Hirose T, et al.
Chemical & Pharmaceutical Bulletin, 30(7), 2399-2409 (1982)
A highly active catalyst for Suzuki-Miyaura cross-coupling reactions of heteroaryl compounds.
Kelvin L Billingsley et al.
Angewandte Chemie (International ed. in English), 45(21), 3484-3488 (2006-04-28)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service