Skip to Content
Merck
All Photos(2)

Key Documents

521795

Sigma-Aldrich

4-Methylphthalonitrile

99%

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
€176.00

€176.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
€176.00

About This Item

Linear Formula:
CH3C6H3-1,2-(CN)2
CAS Number:
Molecular Weight:
142.16
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

€176.00


Check Cart for Availability

Request a Bulk Order

Assay

99%

form

powder or crystals

mp

119-121 °C (lit.)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Cc1ccc(C#N)c(c1)C#N

InChI

1S/C9H6N2/c1-7-2-3-8(5-10)9(4-7)6-11/h2-4H,1H3

InChI key

MDXGRFMFORMPGT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service