Skip to Content
Merck
All Photos(1)

Key Documents

521582

Sigma-Aldrich

1-Chloro-6-iodohexane

96%

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
€113.00

€113.00


Estimated to ship on08 April 2025


Request a Bulk Order

Select a Size

Change View
10 G
€113.00

About This Item

Linear Formula:
I(CH2)6Cl
CAS Number:
Molecular Weight:
246.52
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€113.00


Estimated to ship on08 April 2025


Request a Bulk Order

Quality Level

Assay

96%

refractive index

n20/D 1.5230 (lit.)

bp

110 °C (lit.)

density

1.623 g/mL at 25 °C (lit.)

functional group

chloro
iodo

SMILES string

ClCCCCCCI

InChI

1S/C6H12ClI/c7-5-3-1-2-4-6-8/h1-6H2

InChI key

QTJHNJCILMMRIQ-UHFFFAOYSA-N

General description

1-Chloro-6-iodohexane is an α,ω-dihaloalkane. 1,12-dichlorododecane is formed as a major product from the reduction of 1-chloro-6-iodohexane with nickel(I) salen.[1] It affords 8-chloro-1-octyne on coupling with sodium acetylide.[2]

Application

1-Chloro-6-iodohexane may be used in the preparation of 1,14-dichloro-7-tetradecyne[2] and 10-2′-(methylphenyl)-7-decynyl chloride.[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

New synthesis of meromycolic acid.
Gensler WJ, et al.
The Journal of Organic Chemistry, 44(21), 3643-3652 (1979)
Synthesis of Methyl 11-(2'-Methylphenyl) undecanoate.
Michael WR.
Journal of Chemical and Engineering Data, 11(1), 134-135 (1996)
Homogeneous catalytic reduction of a, ?-dihaloalkanes with electrogenerated nickel (I) salen.
Mubarak MS and Peters DG.
Journal of Electroanalytical Chemistry, 388(1), 195-198 (1995)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service