Skip to Content
Merck
All Photos(1)

Key Documents

521388

Sigma-Aldrich

2,4-Dichlorophenylboronic acid

≥95%

Synonym(s):

2,4-Dichlorobenzeneboronic acid

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€41.60

€41.60


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€41.60

About This Item

Linear Formula:
Cl2C6H3B(OH)2
CAS Number:
Molecular Weight:
190.82
Beilstein:
4983878
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

€41.60


Check Cart for Availability

Request a Bulk Order

Assay

≥95%

mp

246-249 °C (lit.)

functional group

chloro

SMILES string

OB(O)c1ccc(Cl)cc1Cl

InChI

1S/C6H5BCl2O2/c8-4-1-2-5(7(10)11)6(9)3-4/h1-3,10-11H

InChI key

QNEGDGPAXKYZHZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant involved in:
  • Suzuki coupling reactions with alkynyl bromides or aniline / thiophenol
  • Selective hydroxylation to phenols

Reactant involved in synthesis of biologically active molecules including:
  • N-hydroxyindole-2-carboxylates for use as lactate dehydrogenase inhibitors
  • Non-ATP competitive MK2 inhibitors

Other Notes

Contains varying amounts of anhydride

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service