Skip to Content
Merck
All Photos(1)

Key Documents

520497

Sigma-Aldrich

(R)-(−)-p-Toluenesulfinamide

98%

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
€149.00

€149.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
€149.00

About This Item

Linear Formula:
CH3C6H4S(O)NH2
CAS Number:
Molecular Weight:
155.22
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

€149.00


Check Cart for Availability

Request a Bulk Order

Assay

98%

mp

115-118 °C (lit.)

SMILES string

Cc1ccc(cc1)S(N)=O

InChI

1S/C7H9NOS/c1-6-2-4-7(5-3-6)10(8)9/h2-5H,8H2,1H3/t10-/m1/s1

InChI key

YNJDSRPIGAUCEE-SNVBAGLBSA-N

Application

(R)-(-)-p-Toluenesulfinamide may be used in the preparation of E,E-(R)-(-)-N-(2,4-hexadienalidene)-p-toluenesulfinamide by reacting with sorbic aldehyde in the presence of titanium ethoxide.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

d-Amino ?-Keto Esters, a Designed Polyfunctionalized Chiral Building Block for Alkaloid Synthesis. Asymmetric Synthesis of (R)-(+)-2-Phenylpiperidine and (-)-SS20846A.
Davis FA, et al.
Organic Letters, 2(8), 1041-1043 (2000)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service