Skip to Content
Merck
All Photos(1)

Key Documents

520322

Sigma-Aldrich

2-Cyanobenzylzinc bromide solution

0.5 M in THF

Sign Into View Organizational & Contract Pricing

Select a Size

50 ML
€431.00

€431.00


Estimated to ship on01 May 2025


Request a Bulk Order

Select a Size

Change View
50 ML
€431.00

About This Item

Linear Formula:
NCC6H4CH2ZnBr
CAS Number:
Molecular Weight:
261.43
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

€431.00


Estimated to ship on01 May 2025


Request a Bulk Order

Quality Level

concentration

0.5 M in THF

density

0.993 g/mL at 25 °C

functional group

nitrile

storage temp.

2-8°C

SMILES string

Br[Zn]Cc1ccccc1C#N

InChI

1S/C8H6N.BrH.Zn/c1-7-4-2-3-5-8(7)6-9;;/h2-5H,1H2;1H;/q;;+1/p-1

InChI key

LQRNYQAXNZETFP-UHFFFAOYSA-M

Application

2-Cyanobenzylzinc bromide can be used as a reactant:
  • In the metal-catalyzed Negishi cross-coupling reactions to prepare aryl or heteroaryl derivatives via carbon-carbon bond formation.[1]
  • To synthesize 4-(2-Cyanobenzyl)-3′-(trifluoromethyl)biphenyl by reacting with aryl nonaflate in the presence of Pd(dba)2 as a catalyst.[2]
  • To prepare 2-[(2-cyanophenyl)methyl] benzamide by treating with 2-iodobenzamide using a nickel catalyst.[3]

Legal Information

Product of Rieke Metals, Inc.
Rieke is a registered trademark of Rieke Metals, Inc.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Improved nickel-catalyzed cross-coupling reaction conditions between ortho-substituted aryl iodides/nonaflates and alkylzinc iodides in solution and in the solid-phase
Jensen AE, et al.
Tetrahedron, 56(25), 4197-4201 (2000)
Visible-Light-Induced Nickel-Catalyzed Negishi Cross-Couplings by Exogenous-Photosensitizer-Free Photocatalysis
Abdiaj I, et al.
Angewandte Chemie (International Edition in English), 57(28), 8473-8477 (2018)
Palladium-catalyzed cross-coupling reactions with aryl nonaflates: a practical alternative to aryl triflates
Rottlander M and Knochel P
The Journal of Organic Chemistry, 63(1), 203-208 (1998)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service