Skip to Content
Merck
All Photos(1)

Key Documents

519707

Sigma-Aldrich

8-(Tosylamino)quinoline

97%

Synonym(s):

N-(8-Quinolyl)-p-toluenesulfonamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H14N2O2S
CAS Number:
Molecular Weight:
298.36
Beilstein:
264159
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

154-158 °C (lit.)

SMILES string

Cc1ccc(cc1)S(=O)(=O)Nc2cccc3cccnc23

InChI

1S/C16H14N2O2S/c1-12-7-9-14(10-8-12)21(19,20)18-15-6-2-4-13-5-3-11-17-16(13)15/h2-11,18H,1H3

InChI key

ZSMKPYXVUIWTCT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

V D Bovt et al.
Fiziolohichnyi zhurnal (Kiev, Ukraine : 1994), 47(3), 54-57 (2001-08-25)
Alcohol dependence development is accompanied by zinc content decrease in the hippocampus. Young age, repeated physical loads and immobilization are factors, that promote such changes development in more expressed degree.
[Fluorometric method of determining the zinc content in the blood].
A I Merkulov et al.
Laboratornoe delo, (10)(10), 603-604 (1981-01-01)
E D Gol'dberg et al.
Biulleten' eksperimental'noi biologii i meditsiny, 111(2), 135-137 (1991-02-01)
The injections of diabetogenic drugs to rabbits induced a decrease in zinc and calcium content in B insulocytes dependent on the degree of these cells damage and hyperglycemia. The content of these metals in A insulocytes was not significantly altered
J A Lazaris et al.
Endocrinologia experimentalis, 15(2), 99-109 (1981-01-01)
Diabetogenic effects of various analogues of 8-aminoquinoline were studied. Out of 10 compounds investigated only three showed diabetogenic properties: 8-(p-tolylsulfonylamino)-quinoline, 8-(p-benzenesulfonylamino)-quinoline and 8-(p-methylsulfonylamino)-quinoline. After the injection of these compounds a long term persistent hyperglycemia took place in rabbits and also
V A Ieshchenko et al.
Fiziolohichnyi zhurnal (Kiev, Ukraine : 1994), 47(2), 66-69 (2001-06-08)
Zinc content in pancreatic islet cells and hippocampus was studied with the aid of highly sensitive selective cytochemical reaction of 8-(p-toluensulfonylamino)-quinoline. It was shown that the factors stimulating B-insulocyte secretory activity induced the decrease of this metal concentrations in the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service