Skip to Content
Merck
All Photos(2)

Key Documents

518069

Sigma-Aldrich

2-Ethylbenzaldehyde

Synonym(s):

o-Ethylbenzaldehyde

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€305.00

€305.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€305.00

About This Item

Linear Formula:
C2H5C6H4CHO
CAS Number:
Molecular Weight:
134.18
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€305.00


Check Cart for Availability

Request a Bulk Order

refractive index

n20/D 1.538 (lit.)

bp

212 °C (lit.)

density

1.02 g/mL at 25 °C (lit.)

functional group

aldehyde

SMILES string

CCc1ccccc1C=O

InChI

1S/C9H10O/c1-2-8-5-3-4-6-9(8)7-10/h3-7H,2H2,1H3

InChI key

NTWBHJYRDKBGBR-UHFFFAOYSA-N

Application

Reactant for:
  • Preparation of potential antibacterial agents
  • Asymmetric addition reactions
  • Solid-phase synthesis of BODIPY dyes and development of an Ig fluorescent sensor
  • Condensation reactions with phenylenediamine or aminothiophenol catalyzed by Mesoporous mixed metal oxide nanocrystals preparaed from aero gel process

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Directed metalation of aromatic aldimines with lithium 2, 2, 6, 6-tetramethylpiperidide.
Flippin LA, et al.
The Journal of Organic Chemistry, 58(9), 2463-2467 (1993)
Mingzhang Gao et al.
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine, 66(2), 194-202 (2007-10-02)
Small conductance Ca2+-activated K+ (SKCa) channels play an important role in many functions such as neuronal communication and behavioral plasticity, secretion, and cell proliferation. SKCa channel modulation is associated with various brain, heart, and cancer diseases. N-methyl-laudanosine and its structurally
Flavor components of sponge cake and production of 2, 5-dimethyl-4-hydroxy-3 (2H)-furanone in several model systems.
Takei Y.
Agricultural and Biological Chemistry, 41(12), 2361-2368 (1977)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service