Skip to Content
Merck
All Photos(1)

Documents

516562

Sigma-Aldrich

3,5-Difluorophenyl isocyanate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
F2C6H3NCO
CAS Number:
Molecular Weight:
155.10
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

refractive index

n20/D 1.493 (lit.)

bp

159 °C (lit.)

density

1.31 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

Fc1cc(F)cc(c1)N=C=O

InChI

1S/C7H3F2NO/c8-5-1-6(9)3-7(2-5)10-4-11/h1-3H

InChI key

WUCCJYOIAJTFFQ-UHFFFAOYSA-N

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

130.0 °F - closed cup

Flash Point(C)

54.44 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mikhail Soloviev et al.
Journal of nanobiotechnology, 1(1), 4-4 (2003-07-25)
Traditional approaches to protein profiling were built around the concept of investigating one protein at a time and have long since reached their limits of throughput. Here we present a completely new approach for comprehensive compositional analysis of complex protein

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service