Skip to Content
Merck
All Photos(1)

Key Documents

516325

Sigma-Aldrich

4-Methoxy-2-methylbenzaldehyde

95%

Synonym(s):

2-Methyl-p-anisaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3(OCH3)CHO
CAS Number:
Molecular Weight:
150.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

refractive index

n20/D 1.571 (lit.)

bp

262-267 °C (lit.)

density

1.107 g/mL at 25 °C (lit.)

SMILES string

[H]C(=O)c1ccc(OC)cc1C

InChI

1S/C9H10O2/c1-7-5-9(11-2)4-3-8(7)6-10/h3-6H,1-2H3

InChI key

WICYVKGMEJSDAO-UHFFFAOYSA-N

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Compounds of potential interest for the chemotherapy of leprosy.
Buu-Hoi NP, et al.
The Journal of Organic Chemistry, 21(4), 415-418 (1956)
Estrogenic Biphenyls. III. 2-Alkyl-4-methoxybiphenyl-4'-carboxylic Acids.
Sato T and Oki M.
Bulletin of the Chemical Society of Japan, 30(8), 859-862 (1957)
Tamer Awad et al.
Journal of chromatographic science, 45(8), 458-465 (2007-11-21)
The methoxy methyl phenylacetones share an isobaric relationship (equivalent mass but different elemental composition) to the controlled precursor substance 3,4-methylenedioxyphenylacetone. The 10 methoxy methyl phenylacetones as well as the methylenedioxyphenylacetones show essentially equivalent mass spectra with major fragment ions at
Jong H Kim et al.
Annals of clinical microbiology and antimicrobials, 10, 23-23 (2011-06-02)
Disruption of cellular antioxidation systems should be an effective method for control of fungal pathogens. Such disruption can be achieved with redox-active compounds. Natural phenolic compounds can serve as potent redox cyclers that inhibit microbial growth through destabilization of cellular

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service