Skip to Content
Merck
All Photos(2)

Documents

515892

Sigma-Aldrich

Calcium trifluoromethanesulfonate

99.9%

Synonym(s):

Calcium triflate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CF3SO3)2Ca
CAS Number:
Molecular Weight:
338.22
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.22

Assay

99.9%

mp

350 °C (lit.)

SMILES string

[Ca++].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F

InChI

1S/2CHF3O3S.Ca/c2*2-1(3,4)8(5,6)7;/h2*(H,5,6,7);/q;;+2/p-2

InChI key

PUQLFUHLKNBKQQ-UHFFFAOYSA-L

Related Categories

General description

Calcium trifluoromethanesulfonate, also known as calcium triflate [Ca(OTf)2], is widely employed as a Lewis acid catalyst in organic synthesis. It facilitates a range of reactions, including the aldol reaction, Michael additions, and ring-opening reactions of epoxides. In addition, it is also used as a reagent for the preparation of trifluoromethanesulfonate esters, which can be used as leaving groups in a range of organic transformations.

Application

Calcium trifluoromethanesulfonate can be used as a catalyst to synthesize β-amino alcohols via aminolysis of epoxides. It is also used in the one-pot conversion of aldehydes to their corresponding nitrile compounds.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service