Skip to Content
Merck
All Photos(1)

Key Documents

515175

Sigma-Aldrich

Triisopropylsilyl chloride

technical grade, 90%

Synonym(s):

Chlorotriisopropylsilane, TIPSCl, Triisopropylchlorosilane, Triisopropylsilyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)2CH]3SiCl
CAS Number:
Molecular Weight:
192.80
Beilstein:
1737446
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:

grade

technical grade

vapor pressure

1.16 mmHg ( 20 °C)

Assay

90%

refractive index

n20/D 1.452 (lit.)

bp

198 °C/739 mmHg (lit.)

density

0.901 g/mL at 25 °C (lit.)

SMILES string

CC(C)[Si](Cl)(C(C)C)C(C)C

InChI

1S/C9H21ClSi/c1-7(2)11(10,8(3)4)9(5)6/h7-9H,1-6H3

InChI key

KQIADDMXRMTWHZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

147.2 °F - closed cup

Flash Point(C)

64 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service