Skip to Content
Merck
All Photos(1)

Key Documents

513571

Sigma-Aldrich

Luperox® 531M80, 1,1-Bis(tert-amylperoxy)cyclohexane solution

80 wt. % in odorless mineral spirits

Synonym(s):

1,1-Bis(tert-amylperoxy)cyclohexane solution

Sign Into View Organizational & Contract Pricing

Select a Size

500 ML
€103.00

€103.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 ML
€103.00

About This Item

Linear Formula:
C6H10[OOC(CH3)2C2H5]2
CAS Number:
Molecular Weight:
288.42
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

€103.00


Check Cart for Availability

Request a Bulk Order

concentration

80 wt. % in odorless mineral spirits

refractive index

n20/D 1.442

density

0.898 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

CCC(C)(C)OOC1(CCCCC1)OOC(C)(C)CC

InChI

1S/C16H32O4/c1-7-14(3,4)17-19-16(12-10-9-11-13-16)20-18-15(5,6)8-2/h7-13H2,1-6H3

InChI key

IMYCVFRTNVMHAD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Polymerization initiator

Legal Information

Product of Arkema Inc.
Luperox is a registered trademark of Arkema Inc.

Signal Word

Danger

Hazard Classifications

Asp. Tox. 1 - Carc. 1B - Eye Irrit. 2 - Muta. 1B - Org. Perox. C - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.2 - Organic peroxides and self-reacting hazardous materials

WGK

WGK 3

Flash Point(F)

172.4 °F - closed cup

Flash Point(C)

78 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service