Skip to Content
Merck
All Photos(1)

Key Documents

513148

Sigma-Aldrich

(6-Bromohexyloxy)-tert-butyldimethylsilane

99%

Synonym(s):

1-Bromo-6-[(tert-butyldimethylsilyl)oxy]hexane, 6-Bromohexyl tert-butyldimethylsilyl ether, [(6-Bromohexyl)oxy](1,1-dimethylethyl)dimethylsilane

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
€105.00
25 ML
€434.00

€105.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 ML
€105.00
25 ML
€434.00

About This Item

Linear Formula:
(CH3)3CSi(CH3)2O(CH2)6Br
CAS Number:
Molecular Weight:
295.33
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€105.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99%

refractive index

n20/D 1.457 (lit.)

bp

276 °C (lit.)

density

1.053 g/mL at 25 °C (lit.)

functional group

bromo

SMILES string

CC(C)(C)[Si](C)(C)OCCCCCCBr

InChI

1S/C12H27BrOSi/c1-12(2,3)15(4,5)14-11-9-7-6-8-10-13/h6-11H2,1-5H3

InChI key

PBKXRKYUUXKNSL-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Steven McElfresh et al.
Journal of economic entomology, 102(2), 574-584 (2009-05-20)
Marmara gulosa Guillén & Davis (Lepidoptera: Gracillariidae) is a sporadic pest of citrus and a number of other crops in southern and central California. Coupled gas chromatographic-electroantennogram detection analyses of headspace volatiles collected by solid phase microextraction from virgin female
6,6'-Dimethyl-2,2'-bipyridine-4-ester: A pivotal synthon for building tethered bipyridine ligands.
Havas F, et al.
Tetrahedron, 65(36), 7673-7686 (2009)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service