Skip to Content
Merck
All Photos(1)

Key Documents

511781

Sigma-Aldrich

2,6-Difluorophenyl isothiocyanate

98%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€61.30

€61.30


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€61.30

About This Item

Linear Formula:
F2C6H3NCS
CAS Number:
Molecular Weight:
171.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€61.30


Check Cart for Availability

Request a Bulk Order

Assay

98%

refractive index

n20/D 1.604 (lit.)

bp

217 °C (lit.)

density

1.325 g/mL at 25 °C (lit.)

functional group

fluoro
isothiocyanate

storage temp.

2-8°C

SMILES string

Fc1cccc(F)c1N=C=S

InChI

1S/C7H3F2NS/c8-5-2-1-3-6(9)7(5)10-4-11/h1-3H

InChI key

DBSXNGIBAKYMSS-UHFFFAOYSA-N

General description

2,6-Difluorophenyl isothiocyanate (1,3-Difluoro-2-isothiocyanatobenzene), an aryl isocyanate, can be prepared from difluoroaniline.[1] 1H-[1,2,4]triazole-3,5-diamine undergoes acylation with 2,6-difluorophenyl isothiocyanate to form the corresponding 1-acyl-1H-[1,2,4]triazole-3,5-diamine.[2]

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

219.2 °F - closed cup

Flash Point(C)

104 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

1-Acyl-1H-[1,2,4] triazole-3,5-diamine Analogues as Novel and Potent Anticancer Cyclin-Dependent Kinase Inhibitors: Synthesis and Evaluation of Biological Activities.
Lin R, et al.
Journal of Medicinal Chemistry, 48(13), 4208-4211 (2005)
J W Tilley et al.
Journal of medicinal chemistry, 23(12), 1387-1392 (1980-12-01)
Structure-activity studies were carried out on a series of antihypertensive 1-(2-aminoethyl)-3-(substituted phenyl)thioureas. From this class of compounds, the 2,6-dichlorophenyl analogue 2 was found to have potent oral antihypertensive activity in two hypertensive rat models and the renal hypertensive dog. In

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service