All Photos(1)
About This Item
Linear Formula:
Ag3AsO4
CAS Number:
Molecular Weight:
462.52
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
Assay
99.99%
solubility
ammonium hydroxide: slightly soluble(lit.)
density
6.657 g/mL (lit.)
SMILES string
[Ag+].[Ag+].[Ag+].[O-][As]([O-])([O-])=O
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service