Skip to Content
Merck
All Photos(2)

Key Documents

510971

Sigma-Aldrich

(3-Bromophenylethynyl)trimethylsilane

97%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€116.00

€116.00


Estimated to ship on31 May 2025


Request a Bulk Order

Select a Size

Change View
5 G
€116.00

About This Item

Linear Formula:
BrC6H4C≡CSi(CH3)3
CAS Number:
Molecular Weight:
253.21
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€116.00


Estimated to ship on31 May 2025


Request a Bulk Order

Assay

97%

refractive index

n20/D 1.555 (lit.)

bp

80 °C/0.2 mmHg (lit.)

density

1.189 g/mL at 25 °C (lit.)

functional group

bromo

SMILES string

C[Si](C)(C)C#Cc1cccc(Br)c1

InChI

1S/C11H13BrSi/c1-13(2,3)8-7-10-5-4-6-11(12)9-10/h4-6,9H,1-3H3

InChI key

BXXMHQYHYOTTPM-UHFFFAOYSA-N

General description

(3-Bromophenylethynyl)trimethylsilane is an internal alkyne.

Application

(3-Bromophenylethynyl)trimethylsilane may be used in the preparation of 4-(3-bromophenyl)-3-butyn-2-one.[1]

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enantioselective synthesis of both enantiomers of various propargylic alcohols by use of two oxidoreductases.
Schubert T, et al.
European Journal of Organic Chemistry, 2001(22), 4181-4187 (2001)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service