50342
Biphenyl sodium complex solution
technical, 0.5-1.5 M in diethylene glycol diethyl ether
Synonym(s):
Sodium biphenyl complex
About This Item
Recommended Products
grade
technical
Quality Level
form
liquid
shelf life
limited shelf life
reaction suitability
reagent type: reductant
concentration
0.5-1.5 M in diethylene glycol diethyl ether
density
1,100 g/cm3
functional group
phenyl
storage temp.
2-8°C
SMILES string
[Na]c1ccc(cc1)-c2ccccc2
InChI
1S/C12H9.Na/c1-3-7-11(8-4-1)12-9-5-2-6-10-12;/h1,3-10H;
InChI key
UUNVZHGYJSUMDC-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B
Supplementary Hazards
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Does not sustain combustion.
Flash Point(C)
Does not sustain combustion.
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service