Skip to Content
Merck
All Photos(1)

Key Documents

50043

Sigma-Aldrich

Glycerol phosphate calcium salt hydrate

mixture of isomers of α- and β-form, ≥96% (calc. based on dry substance, T)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H7CaO6P · xH2O
CAS Number:
Molecular Weight:
210.14 (anhydrous basis)
Beilstein:
6099192
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Assay

≥96% (calc. based on dry substance, T)

loss

≤15% loss on drying, 150 °C, 4 h

cation traces

Ca: ≥18.3%

storage temp.

2-8°C

SMILES string

O.[Ca++].OCC(O)COP([O-])([O-])=O

InChI

1S/C3H9O6P.Ca.H2O/c4-1-3(5)2-9-10(6,7)8;;/h3-5H,1-2H2,(H2,6,7,8);;1H2/q;+2;/p-2

InChI key

ILSFDSREGJGPRB-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Timothy E L Douglas et al.
International journal of molecular sciences, 21(7) (2020-04-02)
Mineralization of hydrogel biomaterials with calcium phosphate (CaP) is considered advantageous for bone regeneration. Mineralization can be both induced by the enzyme alkaline phosphatase (ALP) and promoted by calcium-binding biomolecules, such as plant-derived polyphenols. In this study, ALP-loaded gellan gum
Grégory Chatelain et al.
Journal of biological inorganic chemistry : JBIC : a publication of the Society of Biological Inorganic Chemistry, 20(3), 497-507 (2014-12-24)
Bone is the main target organ for the storage of several toxic metals, including uranium. But the mode of action of uranium on bones remains poorly understood. To better assess the impact of uranium on bone cells, synthetic biomimetic apatites

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service