Skip to Content
Merck
All Photos(1)

Key Documents

498181

Sigma-Aldrich

2-Chloro-5-iodopyridine

97%

Sign Into View Organizational & Contract Pricing

Select a Size

2 G
€97.60

€97.60


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
2 G
€97.60

About This Item

Empirical Formula (Hill Notation):
C5H3ClIN
CAS Number:
Molecular Weight:
239.44
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€97.60


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

mp

95-98 °C (lit.)

functional group

chloro
iodo

SMILES string

Clc1ccc(I)cn1

InChI

1S/C5H3ClIN/c6-5-2-1-4(7)3-8-5/h1-3H

InChI key

QWLGCWXSNYKKDO-UHFFFAOYSA-N

General description

2-Chloro-5-iodopyridine is a halo-substituted pyridine.

Application

2-Chloro-5-iodopyridine may be used as a reagent in the multi-step synthesis of (±)-epibatidine.[1]
It may be used in the synthesis of:
  • 2-Chloro-5-phenylpyridine via Suzuki coupling reaction with phenylboronic acid dimethyl ester.[2]
  • Exo-5- and exo-6- (6′-chloro-3′-pyridyl)-2-azabicyclo[2.2.1]heptanes via Heck coupling reaction with N-protected 2-azabicyclo[2.2.1]hept-5-enes.[3]
  • Substituted diaryliodonium salts.[4]
  • 3-Exo-5′-(2′-Chloropyridinyl)-8-(ethoxycarbonyl)-8-azabicyclo[3.2.1]octane.[5]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High-yielding one-pot synthesis of diaryliodonium triflates from arenes and iodine or aryl iodides.
Bielawski M and Olofsson B
Chemical Communications (Cambridge, England), 24, 2521-2523 (2007)
A short and efficient total synthesis of (?)-epibatidine.
A short and efficient total synthesis of (?)-epibatidine.
Zhang C and Trudell ML.
The Journal of Organic Chemistry, 61(20), 7189-7191 (1996)
Syntheses of new open-ring and homo-epibatidine analogues from tropinone.
Olivo HF, et al.
The Journal of Organic Chemistry, 64(13), 4966-4968 (1999)
Synthesis of epibatidine isomers: Reductive Heck coupling of 2-azabicyclo [2.2.1] hept-5-ene derivatives.
Cox CD and Malpass JR
Tetrahedron, 55(40), 11879-11888 (1999)
Synthesis of 5-Substituted 2,2'-Bipyridines from Substituted 2-Chloropyridines by a Modified Negishi Cross-Coupling Reaction.
Lutzen A and Hapke M.
European Journal of Organic Chemistry, 2002(14), 2292-2297 (2002)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service