Skip to Content
Merck
All Photos(1)

Key Documents

497592

Sigma-Aldrich

3-Bromo-5-chlorosalicylaldehyde

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H2(Cl)-2-(OH)CHO
CAS Number:
Molecular Weight:
235.46
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

mp

85-88 °C (lit.)

SMILES string

Oc1c(Br)cc(Cl)cc1C=O

InChI

1S/C7H4BrClO2/c8-6-2-5(9)1-4(3-10)7(6)11/h1-3,11H

InChI key

KNOYZLVIXXBBIB-UHFFFAOYSA-N

General description

3-Bromo-5-chlorosalicylaldehyde is a substituted salicyladehyde.

Application

3-Bromo-5-chlorosalicylaldehyde may be used in the synthesis of the following Schiff base compounds:
  • 2-bromo-4-chloro-6-[(E)-o-tolyl­imino­meth­yl]phenol[1]
  • 2-bromo-4-chloro-6-[(1-phenyl­ethyl)­imino­meth­yl]phenol[2]
  • 2-bromo-4-chloro-6-[(E)-(2-chloro­phen­yl)imino­meth­yl]phenol[3]
  • 2′-(3-bromo-5-chloro-2-hydroxy­benzyl­idene)isonicotinohydrazide methanol solvate[4]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

2-Bromo-4-chloro-6-[(1-phenylethyl) iminomethyl] phenol.
Zhang X.
Acta Crystallographica Section E, Structure Reports Online, 64(11), o2084-o2084 (2008)
2-Bromo-4-chloro-6-[(E)-o-tolyliminomethyl] phenol.
Guo JB.
Acta Crystallographica Section E, Structure Reports Online, 66(10), o2689-o2689 (2010)
2-Bromo-4-chloro-6-[(E)-(2-chlorophenyl) iminomethyl] phenol.
Zhang X.
Acta Crystallographica Section E, Structure Reports Online, 65(4), o667-o667 (2009)
2'-(3-Bromo-5-chloro-2-hydroxybenzylidene) isonicotinohydrazide methanol solvate.
Tang CB.
Acta Crystallographica Section E, Structure Reports Online, 64(8), o1381-o1381 (2008)
Yong-Jun Han et al.
Acta chimica Slovenica, 64(1), 179-185 (2017-04-06)
A new cobalt(II) complex, [Co(L1)2(OH2)2] (1), was prepared by the reaction of 3-bromo-5-chlorosalicylaldehyde (HL1) with cobalt nitrate in methanol. Reaction of 1 with cyclopropylamine in methanol afforded the Schiff base cobalt(II) complex, [Co(L2)2] (2), where L2 is the deprotonated form

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service