Skip to Content
Merck
All Photos(1)

Key Documents

495611

Sigma-Aldrich

Ethyl 6-isocyanatohexanoate

98%

Sign Into View Organizational & Contract Pricing

Select a Size

2 G
€89.10

€89.10


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
2 G
€89.10

About This Item

Linear Formula:
OCN(CH2)5CO2C2H5
CAS Number:
Molecular Weight:
185.22
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€89.10


Check Cart for Availability

Request a Bulk Order

Assay

98%

refractive index

n20/D 1.439 (lit.)

bp

253 °C (lit.)

density

1.032 g/mL at 25 °C (lit.)

functional group

amine
ester
isocyanate

storage temp.

2-8°C

SMILES string

CCOC(=O)CCCCCN=C=O

InChI

1S/C9H15NO3/c1-2-13-9(12)6-4-3-5-7-10-8-11/h2-7H2,1H3

InChI key

ZCLBSYCOBSMTMV-UHFFFAOYSA-N

General description

Ethyl 6-isocyanatohexanoate can be synthesized by reacting ethyl 6-aminohexanoate hydrochloride with phosgene.[1] Its enthalpy of vaporization at boiling point has been evaluated.[2]

Application

Ethyl 6-isocyanatohexanoate may be used in the synthesis of 1-(5-ethoxycarbonylpentylcarbamoyl)-5-fluorouracil.[1]

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yaws CL.
Thermophysical Properties of Chemicals and Hydrocarbons, 570-570 (2014)
5-Fluorouracil derivatives. XI. Synthesis of 1-hexylcarbamoyl-5-fluorouracil metabolites.
Ozaki S, et al.
Chemical & Pharmaceutical Bulletin, 34(2), 893-896 (1986)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service