Skip to Content
Merck
All Photos(1)

Key Documents

493473

Sigma-Aldrich

3-Phenoxypropionitrile

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5OCH2CH2CN
CAS Number:
Molecular Weight:
147.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

58-61 °C (lit.)

SMILES string

N#CCCOc1ccccc1

InChI

1S/C9H9NO/c10-7-4-8-11-9-5-2-1-3-6-9/h1-3,5-6H,4,8H2

InChI key

IXAUFLAHUXISCH-UHFFFAOYSA-N

General description

3-Phenoxypropionitrile is an aryl nitrile that can be synthesized by the reaction of acrylonitrile with phenol, catalyzed by (IPr)Cu(OPh) (IPr=1,3-bis(2,6-diisopropylphenyl)imidazol-2-ylidene; Ph=phenyl).[1]

Application

3-Phenoxypropionitrile may be used as a starting material in the synthesis of a phenol novolak-type polymer.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anti-Markovnikov NH and OH additions to electron-deficient olefins catalyzed by well-defined Cu (I) anilido, ethoxide, and phenoxide systems.
Munro-Leighton C, et al.
Journal of the American Chemical Society, 128(5), 1446-1447 (2006)
Formaldehyde polymers, 28. Synthesis of a high molecular weight novolak-type Polymer.
Ninagawa A and Matsuda H.
Makromol. Chem., Rapid Commun., 2(6-7), 449-452 (1981)
Yusuke Yamamoto et al.
The Journal of toxicological sciences, 44(9), 585-600 (2019-09-03)
Amino acid derivative reactivity assay (ADRA) has previously been developed as an alternative method to direct peptide reactivity assay (DPRA) to evaluate key event 1 in skin sensitization mechanisms. However, when using alternative methods for skin sensitization, integrated approaches to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service