Skip to Content
Merck
All Photos(1)

Key Documents

493376

Sigma-Aldrich

p-Toluenesulfonamide-15N

99 atom % 15N

Synonym(s):

p-Tosyl amide-15N

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€828.00

€828.00


Check Cart for Availability

Request more information

Select a Size

Change View
1 G
€828.00

About This Item

Linear Formula:
CH3C6H4SO215NH2
CAS Number:
Molecular Weight:
172.21
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.12

€828.00


Check Cart for Availability

Request more information

isotopic purity

99 atom % 15N

Quality Level

form

solid

mp

138-139 °C (lit.)

mass shift

M+1

SMILES string

Cc1ccc(cc1)S([15NH2])(=O)=O

InChI

1S/C7H9NO2S/c1-6-2-4-7(5-3-6)11(8,9)10/h2-5H,1H3,(H2,8,9,10)/i8+1

InChI key

LMYRWZFENFIFIT-VJJZLTLGSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

395.6 °F - closed cup

Flash Point(C)

202.00 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service