Skip to Content
Merck
All Photos(1)

Key Documents

491993

Sigma-Aldrich

Butyric acid-1,2-13C2

99 atom % 13C

Synonym(s):

Butanoic acid-1,2-13C2

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€379.00

€379.00


Check Cart for Availability

Request more information

Select a Size

Change View
100 MG
€379.00

About This Item

Linear Formula:
CH3CH213CH213CO2H
CAS Number:
Molecular Weight:
90.09
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.12

€379.00


Check Cart for Availability

Request more information

isotopic purity

99 atom % 13C

Quality Level

Assay

99% (CP)

refractive index

n20/D 1.3969 (lit.)

bp

162 °C (lit.)

mp

-6--3 °C (lit.)

density

0.986 g/mL at 25 °C

mass shift

M+2

SMILES string

CC[13CH2][13C](O)=O

InChI

1S/C4H8O2/c1-2-3-4(5)6/h2-3H2,1H3,(H,5,6)/i3+1,4+1

InChI key

FERIUCNNQQJTOY-CQDYUVAPSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

161.6 °F - closed cup

Flash Point(C)

72 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Madeleine Wyss et al.
Frontiers in immunology, 10, 3107-3107 (2020-02-06)
Early life exposure to microbes plays an important role in immune system development. Germ-free mice, or mice colonized with a low-diversity microbiota, exhibit high serum IgE levels. An increase in microbial richness, providing it occurs in a critical developmental window
Adam Shute et al.
Microbiome, 9(1), 186-186 (2021-09-15)
Studies on the inhibition of inflammation by infection with helminth parasites have, until recently, overlooked a key determinant of health: the gut microbiota. Infection with helminths evokes changes in the composition of their host's microbiota: one outcome of which is

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service