Skip to Content
Merck
All Photos(1)

Key Documents

490865

Sigma-Aldrich

Myristic acid-1,2-13C2

99 atom % 13C, 99% (CP)

Synonym(s):

Tetradecanoic acid-1,2-13C2

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
€709.00

€709.00


Check Cart for Availability

Request more information

Select a Size

Change View
500 MG
€709.00

About This Item

Linear Formula:
CH3(CH2)1113CH213CO2H
CAS Number:
Molecular Weight:
230.36
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.12

€709.00


Check Cart for Availability

Request more information

isotopic purity

99 atom % 13C

Assay

99% (CP)

form

solid

bp

250 °C/100 mmHg (lit.)

mp

52-54 °C (lit.)

mass shift

M+2

SMILES string

CCCCCCCCCCCC[13CH2][13C](O)=O

InChI

1S/C14H28O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14(15)16/h2-13H2,1H3,(H,15,16)/i13+1,14+1

InChI key

TUNFSRHWOTWDNC-SBAVNFSASA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service