Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.
490628
Indium(III) nitride
99.9% trace metals basis
Synonym(s):
Indium mononitride, Indium nitride
About This Item
Assay
99.9% trace metals basis
form
powder
reaction suitability
reagent type: catalyst
core: indium
particle size
100 mesh
SMILES string
N#[In]
InChI
1S/In.N
InChI key
NWAIGJYBQQYSPW-UHFFFAOYSA-N
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Conducting polymers such as polyaniline, polythiophene and polyfluorenes are now much in the spotlight for their applications in organic electronics and optoelectronics.
-
What is the Department of Transportation shipping information for this product?
1 answer-
Helpful?
-
-
Is Indium(III) nitride (InN), Product 490628, stable?
1 answer-
Indium nitride is expected to be stable at room temperature, with no other special storage considerations. But, according to the MSDS, it is not compatible with acids and strong oxidants.
Helpful?
-
-
What special property does Indium(III) nitride (InN), Product 490628, have?
1 answer-
It is an inorganic semiconductor.
Helpful?
-
-
What is the band gap of Indium(III) nitride (InN), Product 490628?
1 answer-
Although Sigma-Aldrich has not performed quality control testing for this property, literature suggests that the band gap for InN is expected to be between 0.7 and 0.65 eV, depending on the temperature.
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service