Skip to Content
Merck
All Photos(1)

Key Documents

486167

Sigma-Aldrich

Propionitrile-d5

99 atom % D

Synonym(s):

Pentadeuteriopropionitrile

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€1,930.00

€1,930.00


Estimated to ship on22 May 2025


Request more information

Select a Size

Change View
5 G
€1,930.00

About This Item

Linear Formula:
C2D5CN
CAS Number:
Molecular Weight:
60.11
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.12

€1,930.00


Estimated to ship on22 May 2025


Request more information

isotopic purity

99 atom % D

Assay

99% (CP)

bp

97 °C (lit.)

mp

−93 °C (lit.)

density

0.842 g/mL at 25 °C

mass shift

M+5

SMILES string

[2H]C([2H])([2H])C([2H])([2H])C#N

InChI

1S/C3H5N/c1-2-3-4/h2H2,1H3/i1D3,2D2

InChI key

FVSKHRXBFJPNKK-ZBJDZAJPSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Oral - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

42.8 °F - closed cup

Flash Point(C)

6.00 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service