Skip to Content
Merck
All Photos(1)

Key Documents

483338

Sigma-Aldrich

Cesium triiodide

≥99.9%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€90.60

€90.60


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€90.60

About This Item

Linear Formula:
CsI3
CAS Number:
Molecular Weight:
513.62
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.22

€90.60


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥99.9%

mp

207 °C (lit.)

density

4.51 g/mL at 25 °C (lit.)

SMILES string

I[Cs](I)I

InChI

1S/Cs.I3/c;1-3-2/q+1;-1

InChI key

PSDVEXXHXQVSQT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kathleen Stout et al.
European journal of organic chemistry, 2020(45), 7087-7100 (2021-01-01)
The synthesis and characterization of double porphyrin cage compounds are described. They consist of two porphyrins that are each attached to a diphenylglycoluril-based clip molecule via four ethyleneoxy spacers, and are linked together by a single alkyl chain using "click"-chemistry.
Michaela Rödel et al.
Materials (Basel, Switzerland), 12(1) (2018-12-28)
Dual setting cements composed of an in situ forming hydrogel and a reactive mineral phase combine high compressive strength of the cement with sufficient ductility and bending strength of the polymeric network. Previous studies were focused on the modification with
Subhamoy Saha et al.
The journal of physical chemistry. A, 123(13), 2924-2934 (2019-03-05)
Iodine plays a key role in tropospheric ozone destruction, atmospheric new particle formation, as well as growth. Air-water interface happens to be an important reaction site pertaining to such phenomena. However, except iodide (I-), the behavior of other iodine species
Xianwen Lou et al.
Rapid communications in mass spectrometry : RCM, 30(24), 2628-2634 (2016-09-20)
Ionization in matrix-assisted laser desorption/ionization mass spectrometry (MALDI-MS) is a very complicated process. It has been reported that quaternary ammonium salts show extremely strong matrix and analyte suppression effects which cannot satisfactorily be explained by charge transfer reactions. Further investigation
Gulyaim Sagandykova et al.
International journal of molecular sciences, 21(15) (2020-07-30)
A combination of electrochemistry (EC) and matrix-assisted laser desorption/ionization time-of-flight mass spectrometry (off-line EC-MALDI-TOF-MS) was applied for determination of the studied biologically active compounds (D-glucose, D-fructose, D-galactose, D-pinitol, L-chiro-inositol, and myo-inositol) and their possible electrochemical metabolites. In this work, boron-doped

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service