Skip to Content
Merck
All Photos(1)

Key Documents

482064

Sigma-Aldrich

Poly(dimethylsiloxane), hydride terminated

average Mn ~17,500

Synonym(s):

Hydrogen-terminated di-Me silicones

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€237.00

€237.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
€237.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

€237.00


Check Cart for Availability

Request a Bulk Order

mol wt

average Mn ~17,500

Quality Level

refractive index

n20/D 1.406

viscosity

~500 cSt(lit.)

bp

230 °C (lit.)

density

0.971 g/mL at 25 °C

SMILES string

C[Si](C)=O

InChI

1S/C2H6OSi/c1-4(2)3/h1-2H3

InChI key

SEUDSDUUJXTXSV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

235.0 °F - closed cup

Flash Point(C)

112.8 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

We offer PDMS surface modification kit which allows for durable hydrophilization of various polydimethylsiloxane (PDMS) surfaces.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service