Skip to Content
Merck
All Photos(1)

Key Documents

481424

Sigma-Aldrich

Zinc peroxide

50-60%

Synonym(s):

Zinc dioxide

Sign Into View Organizational & Contract Pricing

Select a Size

1 KG
€112.00

€112.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 KG
€112.00

About This Item

Linear Formula:
ZnO2
CAS Number:
Molecular Weight:
97.39
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23

€112.00


Check Cart for Availability

Request a Bulk Order

form

powder

Quality Level

reaction suitability

reagent type: catalyst
core: zinc

reagent type: oxidant

concentration

50-60%

mp

212 °C (dec.) (lit.)

density

1.57 g/mL at 25 °C (lit.)

SMILES string

[Zn++].[O-][O-]

InChI

1S/O2.Zn/c1-2;/q-2;+2

InChI key

IPTOGCUGCFHDSS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Solutions of zinc peroxide can be prepared from zinc hydroxide precursors, the sol can be subsequently used to prepare zinc nanoparticles.[1][2]

Other Notes

Remainder ZnO

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Ox. Sol. 1 - Skin Irrit. 2

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nonstoichiometric properties of zinc oxide nanoparticles prepared by decomposition of zinc peroxide.
Uekawa N, et al.
Physical Chemistry Chemical Physics, 5(5), 929-934 (2003)
Synthesis of ZnO Nanoparticles by Decomposition of Zinc Peroxide
Uekawa N, et al.
Chemistry Letters (Jpn), 7, 606-607 (2001)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service