Skip to Content
Merck
All Photos(5)

Documents

481394

Sigma-Aldrich

Tetramethyltin

greener alternative

95%

Synonym(s):

Tin tetramethyl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Sn(CH3)4
CAS Number:
Molecular Weight:
178.85
Beilstein:
3647887
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

95%

form

solid

reaction suitability

core: tin

greener alternative product score

old score: 16
new score: 12
Find out more about DOZN™ Scoring

greener alternative product characteristics

Waste Prevention
Safer Solvents and Auxiliaries
Design for Degradation
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

refractive index

n20/D 1.441 (lit.)

bp

74-75 °C (lit.)

mp

−54 °C (lit.)

density

1.291 g/mL at 25 °C (lit.)

greener alternative category

SMILES string

C[Sn](C)(C)C

InChI

1S/4CH3.Sn/h4*1H3;

InChI key

VXKWYPOMXBVZSJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives . This product belongs to category of Re-engineered products, showing key improvements in Green Chemistry Principles “Waste Prevention”, “Safer Solvents and Auxiliaries” and “Design for Degradation”. Click here to view its DOZN scorecard.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

10.4 °F - closed cup

Flash Point(C)

-12 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Florian Dumitrache et al.
Beilstein journal of nanotechnology, 10, 9-21 (2019-01-27)
Zn/F co-doped SnO2 nanoparticles with a mean diameter of less than 15 nm and a narrow size distribution were synthesized by a one-step laser pyrolysis technique using a reactive mixture containing tetramethyltin (SnMe4) and diethylzinc (ZnEt2) vapors, diluted Ar, O2

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service