Skip to Content
Merck
All Photos(1)

Key Documents

481238

Sigma-Aldrich

Bis(methylcyclopentadienyl)zirconium(IV) dichloride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H14Cl2Zr
CAS Number:
Molecular Weight:
320.37
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

Assay

97%

reaction suitability

reagent type: catalyst
core: zirconium

mp

180.5-184 °C (lit.)

SMILES string

Cl[Zr]Cl.C[C]1[CH][CH][CH][CH]1.C[C]2[CH][CH][CH][CH]2

InChI

1S/2C6H7.2ClH.Zr/c2*1-6-4-2-3-5-6;;;/h2*2-5H,1H3;2*1H;/q;;;;+2/p-2

InChI key

LOKCKYUBKHNUCV-UHFFFAOYSA-L

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B - Water-react 2

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service