Skip to Content
Merck
All Photos(1)

Documents

479896

Sigma-Aldrich

Luperox® 231, 1,1-Bis(tert-butylperoxy)-3,3,5-trimethylcyclohexane

92%

Synonym(s):

1,1-Bis(tert-butylperoxy)-3,3,5-trimethylcyclohexane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)3COO]2C6H7(CH3)3
CAS Number:
Molecular Weight:
302.45
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

Assay

92%

refractive index

n20/D 1.441 (lit.)

mp

≥50 °C (SADT) (lit.)

density

0.904 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

CC1CC(C)(C)CC(C1)(OOC(C)(C)C)OOC(C)(C)C

InChI

1S/C17H34O4/c1-13-10-16(8,9)12-17(11-13,20-18-14(2,3)4)21-19-15(5,6)7/h13H,10-12H2,1-9H3

InChI key

NALFRYPTRXKZPN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Polymerization Initiator

Legal Information

Product of Arkema Inc.
Luperox is a registered trademark of Arkema Inc.

Pictograms

Exploding BombFlame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 4 - Org. Perox. B

Storage Class Code

4.1A - Other explosive hazardous materials

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Mitsui et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 31(12), 929-933 (1993-12-01)
1,1-Bis(tert-butylperoxy)-3.3.5-trimethylcyclohexane (BBTC) is widely used in the manufacture of rubber. The present carcinogenicity study in B6C3F1 mice was carried out in order to assess its potential to induce tumours. BBTC was administered at dietary levels of 0 (control), 0.25 and
Iyabo Adekogbe et al.
Biomaterials, 26(35), 7241-7250 (2005-07-14)
Chitosan, the deacetylated derivative of chitin, is a promising scaffold material for skin tissue engineering applications. It is biocompatible and biodegradable, and the degradation products are resorbable. However, the rapid degradation of chitosan and its low mechanical strength are concerns
O I Shadyro et al.
Voprosy meditsinskoi khimii, 43(3), 153-157 (1997-05-01)
The bioantioxidant activity of the synthesized by us on the base of the diatomic phenol compound--3,5-di-t-butylpyrocatechol--has been studied. It was shown that this substance exhibits more pronounced antioxidant properties than tocopherol on the lipid peroxidation process in the rat brain
Xiaopeng Shan et al.
Proceedings of the National Academy of Sciences of the United States of America, 102(15), 5340-5345 (2005-04-02)
The reaction of [Fe(2)(mu-OH)(2)(6-Me(3)-TPA)(2)](2+) (1) [6-Me(3)-TPA, Tris(6-methyl-2-pyridylmethyl)amine] with O(2) in CH(2)Cl(2) at -80 degrees C gives rise to two new intermediates, 2 and 3, before the formation of previously characterized [Fe(2)(O)(O(2))(6-Me(3)-TPA)(2)](2+) (4) that allow the oxygenation reaction to be monitored
M Mitsui et al.
Eisei Shikenjo hokoku. Bulletin of National Institute of Hygienic Sciences, (110)(110), 42-48 (1992-01-01)
A 13-week subchronic toxicity study of 1,1-bis(t-butylperoxy)3,3,5-trimethyl cyclohexane (TMCH) was performed in male and female B6C3F1 mice by feeding a CRF-1 powder diet containing 0, 0.5, 1.0, 2.0 and 4.0% TMCH, to determine the maximum tolerable dose (MTD) for subsequent

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service