Skip to Content
Merck
All Photos(1)

Key Documents

479438

Sigma-Aldrich

Lithium tetramethylcyclopentadienide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LiC5H(CH3)4
CAS Number:
Molecular Weight:
128.14
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

reaction suitability

reagent type: ligand

mp

>350 °C (lit.)

SMILES string

[Li].C[C]1[CH][C](C)[C](C)[C]1C

InChI

1S/C9H13.Li.H/c1-6-5-7(2)9(4)8(6)3;;/h5H,1-4H3;;

InChI key

PTQWRLJKGBNMJV-UHFFFAOYSA-N

Application

Reactant for synthesis of:
  • Cyclopentadienylphosphanes
  • Ansa-cyclopentadienyl-phenoxy titanium(IV) complexes
  • Titanium and zirconium complexes
  • Hafnocene catalysts

Reactant with SiH-functionalized cyclopentadienes

Reactant in the reduction of beta-diketiminato-titanium(III) complexes

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Pyr. Sol. 1

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service