Skip to Content
Merck
All Photos(1)

Key Documents

477788

Sigma-Aldrich

Tetraethyllead solution

50 wt. % in xylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)4Pb
CAS Number:
Molecular Weight:
323.44
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

concentration

50 wt. % in xylene

refractive index

n20/D 1.502

bp

147 °C

density

1.108 g/mL at 25 °C

SMILES string

CC[Pb](CC)(CC)CC

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 3 - Repr. 1A - Skin Irrit. 2 - STOT RE 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system,Liver,Kidney, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

84.2 °F

Flash Point(C)

29 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

EU REACH Annex XIV (Authorisation List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M C Zúñiga et al.
Talanta, 80(2), 504-510 (2009-10-20)
A method for the extraction of triethyl lead (TEL(+)), trimethyl lead (TML(+)), and Pb(2+) from sand was developed using supercritical modified CO(2)-CH(3)OH extraction and in situ complexation with sodium diethyldithiocarbamate (NaDDTC) using a 2(5) factorial exploratory design is described. The
S Ghittori et al.
Giornale italiano di medicina del lavoro ed ergonomia, 27(2), 137-153 (2005-08-30)
The chemical risk in service stations may be due to toxic compounds present in fuel (particularly benzene and additives) and to the emission of exhausts and fine particulate from vehicles. Owing to the elimination of lead (Pb) from fuel and
W F Stewart et al.
Neurology, 66(10), 1476-1484 (2006-05-24)
To determine whether cumulative lead dose in former organolead workers was associated with MRI measures of white matter lesions (WML) and global and structure-specific brain volumes. MRIs, tibia lead, and other measures were obtained from 532 former organolead workers with
Zenon Trela et al.
Comptes rendus biologies, 335(12), 722-730 (2013-01-15)
The patch-clamp technique was used to examine the effect of trimethyllead chloride (Met(3)PbCl) on SV channel activity in red beet (Beta vulgaris L.) taproot vacuoles. It was found that in the control bath the macroscopic currents showed the typical slow
Walter F Stewart et al.
American journal of industrial medicine, 50(10), 729-739 (2007-02-22)
Historically, there has been minimal concern about the effect of adult lead exposure on the brain. Evidence from recent longitudinal studies raise concerns about the long-term effects of past exposure. We initiated three independent longitudinal studies to determine whether cumulative

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service