Skip to Content
Merck
All Photos(1)

Key Documents

47703

Sigma-Aldrich

Fmoc-Glu(OAll)-OH

≥96.0% (HPLC)

Synonym(s):

Fmoc-L-glutamic acid 5-allyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€656.00

€656.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€656.00

About This Item

Empirical Formula (Hill Notation):
C23H23NO6
CAS Number:
Molecular Weight:
409.43
Beilstein:
6670846
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

€656.00


Check Cart for Availability

Request a Bulk Order

Assay

≥96.0% (HPLC)

optical activity

[α]20/D −16.5±2°, c = 1% in DMF

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

SMILES string

OC(=O)[C@H](CCC(=O)OCC=C)NC(=O)OCC1c2ccccc2-c3ccccc13

InChI

1S/C23H23NO6/c1-2-13-29-21(25)12-11-20(22(26)27)24-23(28)30-14-19-17-9-5-3-7-15(17)16-8-4-6-10-18(16)19/h2-10,19-20H,1,11-14H2,(H,24,28)(H,26,27)/t20-/m0/s1

InChI key

LRBARFFNYOKIAX-FQEVSTJZSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ashley E Lepack et al.
Science (New York, N.Y.), 368(6487), 197-201 (2020-04-11)
Vulnerability to relapse during periods of attempted abstinence from cocaine use is hypothesized to result from the rewiring of brain reward circuitries, particularly ventral tegmental area (VTA) dopamine neurons. How cocaine exposures act on midbrain dopamine neurons to precipitate addiction-relevant
Lorna A Farrelly et al.
Nature, 567(7749), 535-539 (2019-03-15)
Chemical modifications of histones can mediate diverse DNA-templated processes, including gene transcription1-3. Here we provide evidence for a class of histone post-translational modification, serotonylation of glutamine, which occurs at position 5 (Q5ser) on histone H3 in organisms that produce serotonin

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service