Skip to Content
Merck
All Photos(1)

Key Documents

475394

Sigma-Aldrich

2-(2-Bromoethoxy)tetrahydro-2H-pyran

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H13BrO2
CAS Number:
Molecular Weight:
209.08
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

contains

~1% K2CO3 as stabilizer

refractive index

n20/D 1.482 (lit.)

bp

62-64 °C/0.4 mmHg (lit.)

density

1.384 g/mL at 25 °C (lit.)

SMILES string

BrCCOC1CCCCO1

InChI

1S/C7H13BrO2/c8-4-6-10-7-3-1-2-5-9-7/h7H,1-6H2

InChI key

GCUOLJOTJRUDIZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-(2-Bromoethoxy)tetrahydro-2H-pyran is an organic building block. It has been synthesized by employing 2-bromoethanol as a starting reagent.[1]

Application

2-(2-Bromoethoxy)tetrahydro-2H-pyran may be used in the synthesis of:
  • 4-(2-chloroethoxy)benzenesulfonyl chloride[2]
  • estrogen ligands bearing carborane[3]
  • 2-(2-(1,3-di((E)-2-(2-(2-(2-(5-(4-(tert-butoxycarbonyl(methyl)amino)styryl)pyridin-2-yloxy)ethoxy)ethoxy)ethoxy)ethoxy)propan-2-yloxy)ethoxy)-tetrahydro-2H-pyran[4]
Used in preparing 2-hydroxyethyl derivatives of cardiolipin analogs,[5] furan-fused compounds,[6] indoles,[7] and pyrazoles.[8]

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

190.4 °F - closed cup

Flash Point(C)

88 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stacy W Remiszewski et al.
Journal of medicinal chemistry, 46(21), 4609-4624 (2003-10-03)
A series of N-hydroxy-3-phenyl-2-propenamides were prepared as novel inhibitors of human histone deacetylase (HDAC). These compounds were potent enzyme inhibitors, having IC(50)s < 400 nM in a partially purified enzyme assay. However, potency in cell growth inhibition assays ranged over
Synlett, 1404-1404 (2006)
Y Endo et al.
Chemistry & biology, 8(4), 341-355 (2001-04-28)
Carboranes (dicarba-closo-dodecaboranes) are a class of carbon-containing polyhedral boron-cluster compounds having remarkable thermal stability and exceptional hydrophobicity. Applications of the unique structural and chemical properties offered by icosahedral carboranes in boron neutron capture therapy have received increasing attention over the
Zhihao Zha et al.
Journal of medicinal chemistry, 54(23), 8085-8098 (2011-10-21)
β-Amyloid plaques (Aβ plaques) in the brain are associated with cerebral amyloid angiopathy (CAA). Imaging agents that could target the Aβ plaques in the living human brain would be potentially valuable as biomarkers in patients with CAA. A new series
Heterocycles, 62, 207-211 (2004)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service